Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium sulfate, 7783-20-2, MFCD00003391, 1kg
Catalog No. 501657242
Encompass
Linear Formula (NH4)2SO4, ≥99.0%, Molecular Weight 132.14, BioXtra
Catalog No. 50-165-7242 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A29391KG
Linear Formula (NH4)2SO4, ≥99.0%, Molecular Weight 132.14, BioXtra