Learn More
Research Products International Corp Ammonium Sulfate, 3 Kilograms
SDP
Catalog No. 501368043
Encompass_Preferred
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.
Catalog No. 50-136-8043 Supplier Research Products International Corp Supplier No. A205103000.0
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.