Learn More
Research Products International Corp Ammonium Sulfate, 5 Kilograms
SDP
Catalog No. 50488486
Encompass_Preferred
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.
Catalog No. 50-488-486 Supplier Research Products International Corp Supplier No. A205105000.0
Add to Cart
Add to Cart
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.