Learn More
Biotang Inc AMMONIUM SULFATE, 500 GM,P-737, CAS # 7783-20-2. Purity > 99%, soluble in water. Shipped at room temperature. Store at room temperature.
Catalog No. 50152333 Shop All BIOTANG Inc Products
Encompass_Preferred
AMMONIUM SULFATE, 500 GM,P-737, CAS # 7783-20-2. Purity > 99%, soluble in water. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-333 Supplier Biotang Inc Supplier No. BTBB879
AMMONIUM SULFATE, 500 GM,P-737, CAS # 7783-20-2. Purity > 99%, soluble in water. Shipped at room temperature. Store at room temperature.