Learn More
Research Products International Corp Ammonium Sulfate, 500 Grams
SDP
Catalog No. 50488485
Encompass_Preferred
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.
Catalog No. 50-488-485 Supplier Research Products International Corp Supplier No. A20510500.0
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.