Learn More
Biotang Inc AMMONIUM SULFATE ~4.1 M, 50 ML, MT-127, ~4.1 M Saturated Ammonium Sulfate solution ((NH4)2SO4). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50152265 Shop All BIOTANG Inc Products
Encompass_Preferred
AMMONIUM SULFATE ~4.1 M, 50 ML, MT-127, ~4.1 M Saturated Ammonium Sulfate solution ((NH4)2SO4). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-265 Supplier Biotang Inc Supplier No. BTBB812
AMMONIUM SULFATE ~4.1 M, 50 ML, MT-127, ~4.1 M Saturated Ammonium Sulfate solution ((NH4)2SO4). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.