Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium sulfate ACS reage5KG
Catalog No. 502903875
Encompass
Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia. It can enhance the phytotoxicity of several herbicides like glyphosates. Ammonium sulfate is formed as a co-product during the manufacture of caprolactam a precursor of nylon.
Catalog No. 50-290-3875 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A49155KG
Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia. It can enhance the phytotoxicity of several herbicides like glyphosates. Ammonium sulfate is formed as a co-product during the manufacture of caprolactam a precursor of nylon.