Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium sulfate, Analytical Standard, Traceable to NIST SRM 194, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111015519 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5519 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5519 Supplier MilliporeSigma Supelco Supplier No. 5984525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Nitrogen Determination According to Kjeldahl Method

TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-20-2
Density 1.77 g/mL (at 25°C)
Physical Form Neat
Linear Formula (NH4)2 SO4
MDL Number MFCD00003391
Melting Point >280°C (decomposition)
Synonym Mascagnite
Quantity 25 g
Formula Weight 132.14
Percent Purity ≥99.5%
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.