Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate Granulated, Supelco

Catalog No. M1173951000 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1173951000 1 kg
M1173955000 5 kg
M1173959025 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1173951000 Supplier MilliporeSigma Supelco Supplier No. 1173951000
Add to Cart
Add to Cart

For Analysis EMSURE™ ACS, ISO, Reag. Ph Eur

  • EMSURE™ granulated salts are designed for improved handling and processing
  • The granulated form provides better flow properties compared to powdered salts, supporting consistent analytical use
  • Produced using a dry granulation process to maintain free-flowing characteristics and stable specifications during storage
  • Reduced dust generation during handling, supporting safer and cleaner use
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Physical Form Granules

Specifications

CAS 7783-20-2
Quantity 1 kg
Percent Purity ≥99.5% (alkalimetric)
Grade ACS
Chemical Name or Material Ammonium Sulfate Granulated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.