Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium sulfate ReagentPl1KG
Catalog No. 502899401
Encompass
Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia. It is reported to enhance the phytotoxicity of several herbicides like glyphosates. It is formed as a co-product during the manufacture of caprolactam a precursor of nylon.
Catalog No. 50-289-9401 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A51321KG
Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia. It is reported to enhance the phytotoxicity of several herbicides like glyphosates. It is formed as a co-product during the manufacture of caprolactam a precursor of nylon.