Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium sulfate ReagentPl5KG
Catalog No. 502903814
Encompass
Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia. It is reported to enhance the phytotoxicity of several herbicides like glyphosates. It is formed as a co-product during the manufacture of caprolactam a precursor of nylon.
Catalog No. 50-290-3814 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A51325KG
Add to Cart
Add to Cart
Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia. It is reported to enhance the phytotoxicity of several herbicides like glyphosates. It is formed as a co-product during the manufacture of caprolactam a precursor of nylon.