Learn More
Strem, An Ascensus Company CAS# 12135-76-1. 100g. Ammonium sulfide, 40-44% aqueous solution. MFCD00010892
SDP
Catalog No. 5090108591
Encompass
CAS# 12135-76-1. 100g. Ammonium sulfide, 40-44% aqueous solution. MFCD00010892. Molecular Weight: 68.14. Molecular Formula: (NH4)2S. Color/form: yellow to dark orange liq. Strem# 02-0950. http://www.strem.com/catalog/v/02-0950/
Catalog No. 50-901-08591 Supplier Strem, An Ascensus Company Supplier No. 020950100G
CAS# 12135-76-1. 100g. Ammonium sulfide, 40-44% aqueous solution. MFCD00010892. Molecular Weight: 68.14. Molecular Formula: (NH4)2S. Color/form: yellow to dark orange liq. Strem# 02-0950. http://www.strem.com/catalog/v/02-0950/