Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium sulfide solution, 12135-76-1, MFCD00010892, 25 mL
Catalog No. NC1510766
Encompass
Linear Formula: (NH4)2S, Molecular Weight: 68.14, 20% in H2O. Ammonium sulfide solution may be used to enhance the performance of type-II InAs/GaSb super lattice photodiodes by surface passivation technique.
Catalog No. NC1510766 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A195225ML
Linear Formula: (NH4)2S, Molecular Weight: 68.14, 20% in H2O. Ammonium sulfide solution may be used to enhance the performance of type-II InAs/GaSb super lattice photodiodes by surface passivation technique.