Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfide Solution (Certified), Fisher Chemical™

Catalog No. p-212744 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
225 mL
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A705-225 225 mL Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A705-225

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

Color Yellow
Assay Percent Range 20 to 24 %
UN Number 2683
Density 1.000g/cm³
Percent Purity 20 to 24%
Grade Certified
Identification Pass Test
Packaging Glass
Vapor Pressure 450mmHg at 20°C
Quantity 225 mL
Physical Form Liquid
Chemical Name or Material Ammonium Sulfide Solution
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes burns by all exposure routes. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:2
Instability:1
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.