Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium sulfide solution1KG
Catalog No. 502899861
Encompass
Ammonium sulfide is an organosulfur compound mainly used as a flavor ingredient due to its characteristic sulfurous odor. The fried bacon aroma produced during the reaction of ammonium sulfide with isovaleraldehyde is due to 2 4 6-triisobutyl-4H-1 3 5-dithiazine and 3 5-diisobutyl-1 2 4-trithiolane formed during the reaction.
Catalog No. 50-289-9861 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2053031KGK
Ammonium sulfide is an organosulfur compound mainly used as a flavor ingredient due to its characteristic sulfurous odor. The fried bacon aroma produced during the reaction of ammonium sulfide with isovaleraldehyde is due to 2 4 6-triisobutyl-4H-1 3 5-dithiazine and 3 5-diisobutyl-1 2 4-trithiolane formed during the reaction.