Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium Sulfite Solution

Catalog No. 1122925L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-229-25L 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 11-229-25L Supplier Sigma Aldrich Supplier No. 112292.5L
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium sulfite is an ammonium salt of sulfurous acid and a byproduct of the flue gas desulfuration process by ammonia. Ammonium sulfite can be oxidized into ammonium sulfate, which can be used as a fertilizer.

Specifications

Melting Point 50°C to 60°C
Molecular Formula C19H34ClN
Linear Formula CH3(CH2)9N(Cl)(CH3)2CH2C6H5
MDL Number MFCD00270130
Quantity 10 g
Molecular Weight (g/mol) 311.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.