Learn More
Medchemexpress LLC Ammonium sulphate | 7783-20-2 | ≥99.0% | 132.14 | 1 G
SDP
Catalog No. 5000365383
Encompass_Preferred
Ammonium sulphate, ≥99.0%, AR is an inorganic sulfate salt primarily utilized in molecular biology. It serves various purposes, including protein precipitation, antibody separation, and enhancing antigen-antibody reactions. Additionally, it functions as a nitrogen source in plant fertilizers. The product exhibits developmental toxicity to freshwater snails but contributes to an increase in Anopheles africanus and Culex mosquito larvae populations in rice paddies. It also inhibits the conversion of glucose to ascorbic acid in plants.
  • Utilized in molecular biology
  • Employed for precipitating proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Serves as a nitrogen source in plant fertilizers
  • Inhibits the conversion of glucose to ascorbic acid in plants
Catalog No. 50-003-65383 Supplier Medchemexpress LLC Supplier No. 1GHYY0261AS1
Ammonium sulphate, ≥99.0%, AR is an inorganic sulfate salt primarily utilized in molecular biology. It serves various purposes, including protein precipitation, antibody separation, and enhancing antigen-antibody reactions. Additionally, it functions as a nitrogen source in plant fertilizers. The product exhibits developmental toxicity to freshwater snails but contributes to an increase in Anopheles africanus and Culex mosquito larvae populations in rice paddies. It also inhibits the conversion of glucose to ascorbic acid in plants.
  • Utilized in molecular biology
  • Employed for precipitating proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Serves as a nitrogen source in plant fertilizers
  • Inhibits the conversion of glucose to ascorbic acid in plants