Learn More
Medchemexpress LLC Ammonium sulphate | 7783-20-2 | 99.3% | 132.14 | 1 G
SDP
Catalog No. 5000365242
Encompass_Preferred
Ammonium sulphate is an inorganic sulfate salt utilized in molecular biology. It can be employed to precipitate proteins, separate antibodies, and enhance antigen-antibody reactions. Additionally, it serves as a nitrogen source in plant fertilizers. For research use only.
  • Precipitates proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Serves as a nitrogen source in plant fertilizers
  • Inhibits the conversion of glucose to ascorbic acid in plants
Catalog No. 50-003-65242 Supplier Medchemexpress LLC Supplier No. 1GHYY0261A
Ammonium sulphate is an inorganic sulfate salt utilized in molecular biology. It can be employed to precipitate proteins, separate antibodies, and enhance antigen-antibody reactions. Additionally, it serves as a nitrogen source in plant fertilizers. For research use only.
  • Precipitates proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Serves as a nitrogen source in plant fertilizers
  • Inhibits the conversion of glucose to ascorbic acid in plants