Learn More
Medchemexpress LLC Ammonium sulphate | 7783-20-2 | 99.3% | 132.14 | 250 MG
SDP
Catalog No. 5000366044
Encompass_Preferred
Ammonium sulphate is an inorganic sulfate salt primarily used in molecular biology. It effectively precipitates proteins, separates antibodies, and enhances antigen-antibody reactions. This compound also functions as a nitrogen source in plant fertilizers and has specific biological activities, including developmental toxicity to freshwater snails and inhibiting glucose conversion to ascorbic acid in plants.
  • Utilized for molecular biology applications
  • Precipitates proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Serves as a nitrogen source in plant fertilizers
  • Exhibits developmental toxicity to freshwater snails
  • Inhibits the conversion of glucose to ascorbic acid in plants
  • Increases Anopheles africanus and Culex mosquito larvae in rice paddies
Catalog No. 50-003-66044 Supplier Medchemexpress LLC Supplier No. 250MGHYY0261A
Ammonium sulphate is an inorganic sulfate salt primarily used in molecular biology. It effectively precipitates proteins, separates antibodies, and enhances antigen-antibody reactions. This compound also functions as a nitrogen source in plant fertilizers and has specific biological activities, including developmental toxicity to freshwater snails and inhibiting glucose conversion to ascorbic acid in plants.
  • Utilized for molecular biology applications
  • Precipitates proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Serves as a nitrogen source in plant fertilizers
  • Exhibits developmental toxicity to freshwater snails
  • Inhibits the conversion of glucose to ascorbic acid in plants
  • Increases Anopheles africanus and Culex mosquito larvae in rice paddies