Learn More
Medchemexpress LLC Ammonium sulphate | 7783-20-2 | 99.3% | 132.14 | 500 MG
SDP
Catalog No. 5000365932
Encompass_Preferred
Ammonium sulphate is an inorganic sulfate salt primarily used in molecular biology. It is effective for protein precipitation, antibody separation, and enhancing antigen-antibody reactions. Additionally, it serves as a nitrogen source in plant fertilizers.
  • Used for molecular biology applications
  • Precipitates proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Acts as a nitrogen source in plant fertilizers
Catalog No. 50-003-65932 Supplier Medchemexpress LLC Supplier No. 500MGHYY0261A
Ammonium sulphate is an inorganic sulfate salt primarily used in molecular biology. It is effective for protein precipitation, antibody separation, and enhancing antigen-antibody reactions. Additionally, it serves as a nitrogen source in plant fertilizers.
  • Used for molecular biology applications
  • Precipitates proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Acts as a nitrogen source in plant fertilizers