Learn More
CTL Scientific Supply Corp AMMONIUM TEST PAPER
Supplier: CTL Scientific Supply Corp 90722
This test paper can be used for the detection of ammonium ions as well as for gaseous ammonia. The white paper turns brownish-yellow in the presence of ammonium ions. Detection limit 10 ppm in solution. 200 strips/box