Learn More
Strem, An Ascensus Company CAS# 12228-87-4. 500g. Ammonium tetraborate tetrahydrate, 99%. MFCD00149601
SDP
Catalog No. 5090113562
Encompass
CAS# 12228-87-4. 500g. Ammonium tetraborate tetrahydrate, 99%. MFCD00149601. Molecular Weight: 263.38. Molecular Formula: (NH4)2B4O7·4H2O. Color/form: colorless xtl. Strem# 93-0203. http://www.strem.com/catalog/v/93-0203/
Catalog No. 50-901-13562 Supplier Strem, An Ascensus Company Supplier No. 930203500G
CAS# 12228-87-4. 500g. Ammonium tetraborate tetrahydrate, 99%. MFCD00149601. Molecular Weight: 263.38. Molecular Formula: (NH4)2B4O7·4H2O. Color/form: colorless xtl. Strem# 93-0203. http://www.strem.com/catalog/v/93-0203/