Learn More
Strem, An Ascensus Company CAS# 13820-40-1. 5g. Ammonium tetrachloropalladate(II) (99.998%-Pd) PURATREM. MFCD00010884
SDP
Catalog No. 5090108596
Encompass
CAS# 13820-40-1. 5g. Ammonium tetrachloropalladate(II) (99.998%-Pd) PURATREM. MFCD00010884. Molecular Weight: 284.29. Molecular Formula: (NH4)2PdCl4. Color/form: red to brown pwdr. Strem# 02-1275. http://www.strem.com/catalog/v/02-1275/
Catalog No. 50-901-08596 Supplier Strem, An Ascensus Company Supplier No. 0212755G
CAS# 13820-40-1. 5g. Ammonium tetrachloropalladate(II) (99.998%-Pd) PURATREM. MFCD00010884. Molecular Weight: 284.29. Molecular Formula: (NH4)2PdCl4. Color/form: red to brown pwdr. Strem# 02-1275. http://www.strem.com/catalog/v/02-1275/