Ammonium tetrathiomolybdate, 99.95% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4349314

Quantity 25g

 View more versions of this product

Catalog No. AA4349314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15060-55-6
    Physical Form Crystalline
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula H8MoN2S4
    Formula Weight 260.27
    MDL Number MFCD00136013
    Synonym Ammonium molybdenum sulfide
    Chemical Name or Material Ammonium tetrathiomolybdate

    Specifications

    Formula Weight 260.27
    Quantity 25g
    Physical Form Crystalline
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula H8MoN2S4
    MDL Number MFCD00136013
    CAS 15060-55-6
    Synonym Ammonium molybdenum sulfide
    Chemical Name or Material Ammonium tetrathiomolybdate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit