Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tetrathiomolybdate, 99.95%, (trace metal basis)

Catalog No. AC389530500
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
Packaging:
Glass bottle

CAS: 15060-55-6 | H10MoN2S4 | 262.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15060-55-6
Molecular Formula H10MoN2S4
Molecular Weight (g/mol) 262.28
MDL Number MFCD00136013
InChI Key ZKKLPDLKUGTPME-UHFFFAOYSA-N
Synonym ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4
PubChem CID 15251598
SMILES [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S

Specifications

Melting Point 300.0°C
Color White
Physical Form Crystals or Powder
Quantity 50 g
Assay Percent Range 99.95%
Linear Formula (NH4)2MoS4
Merck Index 15,557
IUPAC Name diazanium;bis(sulfanylidene)molybdenum;sulfanide
Formula Weight 260.28
Percent Purity 99.95%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ammonium tetrathiomolybdate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.