Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium tetrathiotungstate

Catalog No. 33673425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-673-425G 25 g
33-673-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-673-425G Supplier Sigma Aldrich Supplier No. 33673425G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This classic building block for transition metal sulfide cluster compounds was recently used in a low-temperature, solid state synthesis of a new series of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands.

Specifications

CAS 13862-78-7
Quantity 25 g
Molecular Formula H8N2S4W
Linear Formula (NH4)2 WS4
MDL Number MFCD00061410
Synonym Ammonium tungsten sulfide ((NH4)2 WS4); Bis(ammonium) tetrathiotungstate(2-) ; Diammonium tetrathiotungstate; Diammonium tetrathiotungstate(2-)
Molecular Weight (g/mol) 348.18
Percent Purity ≥99.9% trace metals basis
Density 2.71 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.