Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Thiosulfate, Granular, Purified, Spectrum™ Chemical
SDP

Catalog No. 18608807 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-807 500 g Poly Bottle
18-608-808 2.5 kg Poly Bottle
18-608-809 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-608-807 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1273500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7783-18-8

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 500 g
Molecular Weight (g/mol) 148.21
Grade Purified
Packaging Poly Bottle
Chemical Name or Material Ammonium Thiosulfate, Granular

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.