Learn More
Medchemexpress LLC Ammonium trifluoroac 100g
SDP
Supplier:  Medchemexpress LLC HYW105853100G
Encompass_Preferred
Ammonium trifluoroacetate is an additive for regulating and calibrating new liquid/mass spectrometry systems[1]
Catalog No. 50-002-06474