Learn More
Medchemexpress LLC Ammonium trifluoroac 50g
SDP
Catalog No. 5000206097
Encompass_Preferred
Ammonium trifluoroacetate is an additive for regulating and calibrating new liquid/mass spectrometry systems[1]
Catalog No. 50-002-06097 Supplier Medchemexpress LLC Supplier No. HYW10585350G
Ammonium trifluoroacetate is an additive for regulating and calibrating new liquid/mass spectrometry systems[1]