Learn More
Aviva Systems Biology Corp AMN Antibody - N-terminal region (ARP49982_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49982_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMN. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human AMN. Alternate names: PRO1028ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-476