Learn More
Cayman Chemical Amodiaquine, Purity: ≥95%, CAS: 86-42-0, 50 mg
SDP
Catalog No. 502973289
Encompass_Preferred
A prodrug form of N-desethyl amodiaquine; active against several strains of P. falciparum in vitro (EC50s = 0.23-0.52 nM) and exhibits a synergistic effect when used in combination with N-desethyl amodiaquine; dose-dependently inhibits development of parasitemia in a mouse model of P. berghei infection
Catalog No. 50-297-3289 Supplier Cayman Chemical Supplier No. 1595450 MG
A prodrug form of N-desethyl amodiaquine; active against several strains of P. falciparum in vitro (EC50s = 0.23-0.52 nM) and exhibits a synergistic effect when used in combination with N-desethyl amodiaquine; dose-dependently inhibits development of parasitemia in a mouse model of P. berghei infection