Learn More
Apexbio Technology LLC Amodiaquine dihydrochloride dihydrate 6398-98-7 500mg
SDP
Catalog No. 5000090848
Encompass_Preferred
Amodiaquine dihydrochloride dihydrate (CAS 6398-98-7) is a small-molecule inhibitor targeting histamine N-methyltransferase (HNMT) It is designed to inhibit HNMT thereby affecting intracellular histamine metabolism Amodiaquine dihydrochloride dihydrate exerts its biological activity primarily through inhibition of HNMT In biochemical assays involving rat kidney-derived HNMT amodiaquine demonstrates inhibitory activity with a reported Ki value of approximately 18 6 nM Based on these pharmacological properties amodiaquine dihydrochloride dihydrate holds research potential in investigating histamine metabolism pathways in inflammatory disease models
Catalog No. 50-000-90848 Supplier Apexbio Technology LLC Supplier No. B6134500
Amodiaquine dihydrochloride dihydrate (CAS 6398-98-7) is a small-molecule inhibitor targeting histamine N-methyltransferase (HNMT) It is designed to inhibit HNMT thereby affecting intracellular histamine metabolism Amodiaquine dihydrochloride dihydrate exerts its biological activity primarily through inhibition of HNMT In biochemical assays involving rat kidney-derived HNMT amodiaquine demonstrates inhibitory activity with a reported Ki value of approximately 18 6 nM Based on these pharmacological properties amodiaquine dihydrochloride dihydrate holds research potential in investigating histamine metabolism pathways in inflammatory disease models