Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amodiaquine impurity 1 | 81099-86-7 | 270.72 | 500 MG
SDP

Catalog No. 5000451278
Encompass_Preferred

Amodiaquine impurity 1 is a chemical identified as an impurity of Amodiaquine. It is intended for research use only by experienced personnel in laboratory settings for the manufacture of substances. This product has not been fully validated for medical applications.

  • Store at 4°C, protected from light, and under nitrogen.
  • In solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light and under nitrogen.
  • Soluble in DMSO at 20 mg/mL (73.88 mM) with ultrasonic, warming, and heating to 60°C.

Catalog No. 50-004-51278 Supplier Medchemexpress LLC Supplier No. HYW008727500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amodiaquine impurity 1 is a chemical identified as an impurity of Amodiaquine. It is intended for research use only by experienced personnel in laboratory settings for the manufacture of substances. This product has not been fully validated for medical applications.

  • Store at 4°C, protected from light, and under nitrogen.
  • In solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light and under nitrogen.
  • Soluble in DMSO at 20 mg/mL (73.88 mM) with ultrasonic, warming, and heating to 60°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.