Learn More
Medchemexpress LLC HY-10982 5mg Medchemexpress, Amonafide CAS:69408-81-7 Purity:>98%
SDP
Catalog No. 501968722
Encompass_Preferred
Medchemexpress, HY-10982 5mg Amonafide CAS:69408-81-7 Amonafide is a topoisomerase II inhibitor and DNA intercalator that induces apoptotic signaling by blocking the binding of Topo II to DNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8722 Supplier Medchemexpress LLC Supplier No. HY109825MG
Medchemexpress, HY-10982 5mg Amonafide CAS:69408-81-7 Amonafide is a topoisomerase II inhibitor and DNA intercalator that induces apoptotic signaling by blocking the binding of Topo II to DNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.