Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Ammonium Chloride | 12125-02-9 | MFCD00011420 | 1 L
SDP

Catalog No. NC0763752
Encompass_Preferred

Ammonium Chloride | Mol Wt: 53.49 | 12125-02-9 | MFCD00011420 | 1 L

Catalog No. NC0763752 Supplier Aqua Solutions Supplier No. 04251L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Chloride | Mol Wt: 53.49 | 12125-02-9 | MFCD00011420 | 1 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.