Learn More
AdipoGen Amorfrutin A (500 µg)
SDP
Supplier:  AdipoGen AGCN20462C500
Encompass_Preferred
Chemical. CAS: 80489-90-3. Formula: C21H24O4. MW: 340.4. Isolated from Amorpha fruticosa. Antidiabetic agent. Potent and selective partial agonist of peroxisome proliferator-activated receptor gamma (PPARgamma), a nuclear receptor regulating lipid and glucose metabolism. Has nanomolar binding affinity to PPARgamma and micromolar binding to the isotypes PPARalpha and PPARbeta/delta. Glucose-lowering agent. Improves insulin sensitivity, glucose tolerance and blood lipid variables without increase of fat storage or hepatoxicity. Shows potent PPARgamma-dependent anti-inflammatory activity. Potent NF-kappaB activation inhibitor by the suppression of IkappaBalpha degradation, p65 nuclear translocation and DNA-binding activity. Antimicrobial agent.
Catalog No. 50-205-3635