Learn More
AdipoGen Amorfrutin B (1 mg)
SDP
Supplier:  AdipoGen AGCN20464M001
Encompass_Preferred
Chemical. CAS: 78916-42-4 and 1174387-94-0. Formula: C26H32O4. MW: 408.5. Isolated from Amorpha fruticosa. Antidiabetic agent. Potent and selective partial agonist of peroxisome proliferator-activated receptor gamma (PPARgamma), a nuclear receptor regulating lipid and glucose metabolism. Has low nanomolar binding affinity to PPARgamma and micromolar binding to the isotypes PPARalpha and PPARbeta/delta. The binding affinity constant to PPARgamma is 12 times lower than Amorfrutin A (Prod. No. AG-CN2-0462). Glucose-lowering agent. Improves insulin sensitivity, glucose tolerance and blood lipid variables without increase of fat storage or hepatoxicity. Shows potent PPARgamma-dependent anti-inflammatory activity. Antimicrobial agent.
Catalog No. 50-205-3639