Learn More
AdipoGen Amorolfine hydrochloride (500 mg)
SDP
Supplier:  AdipoGen CDXA0301M500
Encompass_Preferred
Chemical. CAS: 78613-38-4. Formula: C21H35ON . HCl. MW: 317.51 . 36.46. Amorolfine (or amorolfin), is a morpholine antifungal drug that inhibits D14-sterol reductase and cholestenol D7-D8 isomerase, consequently depleting ergosterol and causes ignosterol to accumulate in the fungal cytoplasmic cell membranes in vivo. It acts primarily by inhibiting ergosterol biosynthesis, a component of fungal cell membrane, and possesses both fungistatic and fungicidal activity. Its spectrum of in vitro activity includes dermatophyte, dimorphic, dematiaceous and filamentous fungi and some yeasts.
Catalog No. 50-205-4264