Learn More
Apexbio Technology LLC Amorolfine HCl 78613-38-4 200mg
SDP
Supplier:  Apexbio Technology LLC B2077200
Encompass_Preferred
Amorolfine hydrochloride (78613-38-4) is a small-molecule inhibitor targeting 14-reductase and 7 8-isomerase enzymes essential for fungal ergosterol synthesis It is designed to disrupt fungal sterol biosynthesis thereby impairing membrane stability and fungal cell viability Amorolfine hydrochloride exerts its biological activity primarily through inhibition of 14-reductase and 7 8-isomerase In in vitro studies Amorolfine hydrochloride demonstrates antifungal activity with IC50 values ranging from approximately 0 02 to 0 5 g/mL depending on fungal species and experimental conditions Based on these pharmacological properties Amorolfine hydrochloride holds research potential in investigating antifungal activity against fungal pathogens including models of human nail infection
Catalog No. 50-000-89193