Learn More
Medchemexpress LLC Amorolfine hydrochloride | 78613-38-4 | 99.6% | 200 MG
SDP
Catalog No. 5000405588
Encompass_Preferred
Amorolfine hydrochloride (Standard) is an analytical standard and a potent anti-fungal agent that inhibits ergosterol biosynthesis. It is intended for research and analytical applications, with potential for the research of Neoscytalidium dimidiatum onychomycosis.
  • Inhibits ergosterol biosynthesis
  • Possesses fungistatic and fungicidal activity
  • Effective against dermatophyte fungi, filamentous fungi, pathogenic yeasts, dimorphic fungi, and dematiaceous fungi
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-05588 Supplier Medchemexpress LLC Supplier No. HYB0238200MG
Amorolfine hydrochloride (Standard) is an analytical standard and a potent anti-fungal agent that inhibits ergosterol biosynthesis. It is intended for research and analytical applications, with potential for the research of Neoscytalidium dimidiatum onychomycosis.
  • Inhibits ergosterol biosynthesis
  • Possesses fungistatic and fungicidal activity
  • Effective against dermatophyte fungi, filamentous fungi, pathogenic yeasts, dimorphic fungi, and dematiaceous fungi
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS