Learn More
Medchemexpress LLC Amorolfine (hydrochloride) | 78613-38-4 | 99.8% | 500 MG
SDP
Catalog No. 5000406928
Encompass_Preferred
Amorolfine hydrochloride is a potent anti-fungal agent that inhibits ergosterol biosynthesis. It has potential for research on *Neoscytalidium dimidiatum* onychomycosis, exhibiting both fungistatic and fungicidal activity.
  • Potent anti-fungal agent
  • Inhibits ergosterol biosynthesis
  • Exhibits fungistatic and fungicidal activity
  • Available as a white to off-white solid
  • Store at 4°C, sealed, away from moisture
Catalog No. 50-004-06928 Supplier Medchemexpress LLC Supplier No. HYB0238500MG
Add to Cart
Add to Cart
Amorolfine hydrochloride is a potent anti-fungal agent that inhibits ergosterol biosynthesis. It has potential for research on *Neoscytalidium dimidiatum* onychomycosis, exhibiting both fungistatic and fungicidal activity.
  • Potent anti-fungal agent
  • Inhibits ergosterol biosynthesis
  • Exhibits fungistatic and fungicidal activity
  • Available as a white to off-white solid
  • Store at 4°C, sealed, away from moisture