Learn More
Medchemexpress LLC Amorolfine hydrochloride | 78613-38-4 | MFCD00903738 | 99.8% | 1 ML
SDP
Catalog No. 5000406890
Encompass_Preferred
Amorolfine hydrochloride (Ro 14-4767/002) is a potent antifungal agent that inhibits ergosterol biosynthesis, a crucial component of fungal cell membranes. This mechanism of action makes it effective against a broad spectrum of fungi. It holds potential for research into *Neoscytalidium dimidiatum* onychomycosis and other fungal infections.
  • Potent antifungal agent
  • Inhibits ergosterol biosynthesis
  • Effective against a broad spectrum of fungi
  • Potential for research into onychomycosis
Catalog No. 50-004-06890 Supplier Medchemexpress LLC Supplier No. HYB023810MM/1ML
Amorolfine hydrochloride (Ro 14-4767/002) is a potent antifungal agent that inhibits ergosterol biosynthesis, a crucial component of fungal cell membranes. This mechanism of action makes it effective against a broad spectrum of fungi. It holds potential for research into *Neoscytalidium dimidiatum* onychomycosis and other fungal infections.
  • Potent antifungal agent
  • Inhibits ergosterol biosynthesis
  • Effective against a broad spectrum of fungi
  • Potential for research into onychomycosis