Learn More
Medchemexpress LLC Amorolfine hydrochloride (Standard) | 78613-38-4 | 99.6% | 100 MG
SDP
Catalog No. 5000407392
Encompass_Preferred
Amorolfine hydrochloride (Standard) is the analytical standard of Amorolfine hydrochloride and is intended for research and analytical applications. Amorolfine hydrochloride is a potent anti-fungal agent that inhibits ergosterol biosynthesis. It has potential for research into Neoscytalidium dimidiatum onychomycosis. The compound is an analytical standard used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Amorolfine hydrochloride.
  • Intended for research and analytical applications.
  • Potent anti-fungal agent.
  • Inhibits ergosterol biosynthesis.
  • Potential for research of Neoscytalidium dimidiatum onychomycosis.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-07392 Supplier Medchemexpress LLC Supplier No. HYB0238R100MG
Amorolfine hydrochloride (Standard) is the analytical standard of Amorolfine hydrochloride and is intended for research and analytical applications. Amorolfine hydrochloride is a potent anti-fungal agent that inhibits ergosterol biosynthesis. It has potential for research into Neoscytalidium dimidiatum onychomycosis. The compound is an analytical standard used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Amorolfine hydrochloride.
  • Intended for research and analytical applications.
  • Potent anti-fungal agent.
  • Inhibits ergosterol biosynthesis.
  • Potential for research of Neoscytalidium dimidiatum onychomycosis.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.