Learn More
Medchemexpress LLC Amorolfine hydrochloride (Standard) | 78613-38-4 | 99.6% | 25 MG
SDP
Catalog No. 5000450648
Encompass_Preferred
Amorolfine hydrochloride (Standard) is an analytical standard and a potent anti-fungal agent that inhibits ergosterol biosynthesis. This product is intended for research and analytical applications, and has potential for the study of *Neoscytalidium dimidiatum* onychomycosis. It acts primarily by inhibiting ergosterol biosynthesis, a component of fungal cell membranes, and and possesses both fungistatic and fungicidal activity.
  • Analytical standard for reference assays
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Suitable for HPLC applications
  • Suitable for GC applications
  • Suitable for MS applications
Catalog No. 50-004-50648 Supplier Medchemexpress LLC Supplier No. 25MGHYB0238R
Amorolfine hydrochloride (Standard) is an analytical standard and a potent anti-fungal agent that inhibits ergosterol biosynthesis. This product is intended for research and analytical applications, and has potential for the study of *Neoscytalidium dimidiatum* onychomycosis. It acts primarily by inhibiting ergosterol biosynthesis, a component of fungal cell membranes, and and possesses both fungistatic and fungicidal activity.
  • Analytical standard for reference assays
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Suitable for HPLC applications
  • Suitable for GC applications
  • Suitable for MS applications