Learn More
Medchemexpress LLC Amoscanate | 26328-53-0 | 99.5% | 10 MG
SDP
Catalog No. 5000340677
Encompass_Preferred
Amoscanate (CGP4540) is a phenyl isothiocyanate where a 4-nitroanilinyl group replaces hydrogen at the para-position. It acts as an anti-schistosomal agent and uncoupler of oxidative phosphorylation, known to potently injure rodent ependyma. This compound is intended for research use only.
  • Effective against schistosomiasis
  • Classified as an isothiocyanate compound
  • Acts by uncoupling oxidative phosphorylation
  • Has a purity of 99.5%
Catalog No. 50-003-40677 Supplier Medchemexpress LLC Supplier No. HY12905110MG
Amoscanate (CGP4540) is a phenyl isothiocyanate where a 4-nitroanilinyl group replaces hydrogen at the para-position. It acts as an anti-schistosomal agent and uncoupler of oxidative phosphorylation, known to potently injure rodent ependyma. This compound is intended for research use only.
  • Effective against schistosomiasis
  • Classified as an isothiocyanate compound
  • Acts by uncoupling oxidative phosphorylation
  • Has a purity of 99.5%