Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amoscanate | 26328-53-0 | 99.5% | 50 MG
SDP

Catalog No. 5000338306
Encompass_Preferred

Amoscanate (CGP4540) is a phenyl isothiocyanate that functions as an anti-schistosomal agent. As an isothiocyanate compound and an uncoupler of oxidative phosphorylation, it has been shown to potently injure rodent ependyma. It appears as a yellow to orange solid.

  • Acts as an anti-schistosomal agent
  • Functions as an isothiocyanate compound
  • Uncoupler of oxidative phosphorylation
  • Molecular formula: C13H9N3O2S
  • Molecular weight: 271.29
  • Solid appearance
  • Yellow to orange color
  • Soluble in DMSO
  • Targets parasite and oxidative phosphorylation
  • Involved in anti-infection and metabolic enzyme/protease pathways

Catalog No. 50-003-38306 Supplier Medchemexpress LLC Supplier No. HY12905150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amoscanate (CGP4540) is a phenyl isothiocyanate that functions as an anti-schistosomal agent. As an isothiocyanate compound and an uncoupler of oxidative phosphorylation, it has been shown to potently injure rodent ependyma. It appears as a yellow to orange solid.

  • Acts as an anti-schistosomal agent
  • Functions as an isothiocyanate compound
  • Uncoupler of oxidative phosphorylation
  • Molecular formula: C13H9N3O2S
  • Molecular weight: 271.29
  • Solid appearance
  • Yellow to orange color
  • Soluble in DMSO
  • Targets parasite and oxidative phosphorylation
  • Involved in anti-infection and metabolic enzyme/protease pathways

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.