Learn More
Medchemexpress LLC HY-129051 5mg , Amoscanate CAS:26328-53-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1290515MG
Encompass_Preferred
HY-129051 5mg Amoscanate CAS: 26328-53-0 Amoscanate (cgp4540) is phenyl isothiocyanate in which the hydrogen at the para-position has been replaced by a 4-nitroanilinyl group. Amoscanate is an anti-schistosomal agent. Amoscanate, as an isothiocyanate compound and uncoupler of oxidative phosphorylation, potently injures rodent ependyma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1742