Learn More
Medchemexpress LLC Amotosalen hydrochloride | 161262-45-9 | MFCD09970539 | 99.9% | 337.8 g/mol | C17H20ClNO4 | 1 ML
SDP
Catalog No. 5000239955
Encompass_Preferred
Amotosalen hydrochloride is a light-activated psoralen compound that crosslinks DNA and RNA upon UVA exposure, used for pathogen inactivation studies and for research applications requiring covalent nucleic acid modification. Supplied as a 10 mM solution (1 mL) with high purity and documented handling information, it is intended for laboratory research use under appropriate safety protocols.
  • Photoactivated crosslinking of DNA and RNA for pathogen inactivation studies.
  • Supplied as a ready-to-use 10 mM solution for consistent dosing.
  • High purity suitable for analytical and biological research applications.
  • Well-characterized molecular properties for reproducible results.
  • Includes SDS and datasheet documentation for safe handling and use.
Catalog No. 50-002-39955 Supplier Medchemexpress LLC Supplier No. HY107004A10MM 1ML
Amotosalen hydrochloride is a light-activated psoralen compound that crosslinks DNA and RNA upon UVA exposure, used for pathogen inactivation studies and for research applications requiring covalent nucleic acid modification. Supplied as a 10 mM solution (1 mL) with high purity and documented handling information, it is intended for laboratory research use under appropriate safety protocols.
  • Photoactivated crosslinking of DNA and RNA for pathogen inactivation studies.
  • Supplied as a ready-to-use 10 mM solution for consistent dosing.
  • High purity suitable for analytical and biological research applications.
  • Well-characterized molecular properties for reproducible results.
  • Includes SDS and datasheet documentation for safe handling and use.