Learn More
Apexbio Technology LLC Amoxapine 14028-44-5 250mg
SDP
Catalog No. 5000088266
Encompass_Preferred
Amoxapine is a small-molecule inhibitor targeting glycine transporter 2a (GLYT2a) It is designed to inhibit synaptic glycine reuptake thereby modulating glycinergic neurotransmission pathways Amoxapine exerts its biological activity primarily through inhibition of GLYT2a It demonstrates this inhibitory activity with an IC50 value of approximately 92 M Based on these pharmacological properties amoxapine holds research potential in studies of glycine-mediated synaptic transmission characterization of neurological conditions and exploration of therapeutic approaches targeting glycine transporter systems
Catalog No. 50-000-88266 Supplier Apexbio Technology LLC Supplier No. B1660250
Amoxapine is a small-molecule inhibitor targeting glycine transporter 2a (GLYT2a) It is designed to inhibit synaptic glycine reuptake thereby modulating glycinergic neurotransmission pathways Amoxapine exerts its biological activity primarily through inhibition of GLYT2a It demonstrates this inhibitory activity with an IC50 value of approximately 92 M Based on these pharmacological properties amoxapine holds research potential in studies of glycine-mediated synaptic transmission characterization of neurological conditions and exploration of therapeutic approaches targeting glycine transporter systems