Learn More
Cayman Chemical AmoxIcIlIn-d4 500ug
SDP
Supplier:  Cayman Chemical 25428500 UG
Encompass_Preferred
An internal standard for the quantification of amoxicillin by GC- or LC-MS
Catalog No. 50-293-6043